Microplates, NeutrAvidin™ coated, 96- and 384- well, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
15129 15117 15128 15116 15123 15127 15400 15401 15402 15507 15508 15509 15510 15511 15513 15216 15217
PIER15129EA 253 EUR
PIER15129 PIER15117 PIER15128 PIER15116 PIER15123 PIER15127 PIER15400 PIER15401 PIER15402 PIER15507 PIER15508 PIER15509 PIER15510 PIER15511 PIER15513 PIER15216 PIER15217
Microplates, NeutrAvidin™ coated, 96- and 384- well, Pierce™
Microplaten Immunologie-microplaten en -strips
PS, NeutrAvidin™ coated plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
  • Low nonspecific binding
  • Easy and gentle immobilisation of biotin-containing conjugates
  • No denaturing of the protein component of a conjugate upon binding
  • Pre-blocked with Blocker™ BSA or SuperBlock™ blocking buffer and ready-to-use
  • High binding capacity plates provides wider detection range and better curve linearity for small biotinylated ligands
Clear plates for colorimetric ELISA and other assays. White plates forMore Product Information