Immunoassay microplates, anti-GST coated, 96-well strip plates, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
15144
PIER15144EA 275 EUR
PIER15144
Immunoassay microplates, anti-GST coated, 96-well strip plates, Pierce™
Microplaten Immunologie-microplaten en -strips
Pierce™ Anti-GST coated plates are polystyrene microplates coated with mouse monoclonal anti-GST antibody and pre-blocked for immediate use.
  • Antibody-based - plates are coated with mouse monoclonal anti-GST antibody
  • Sensitive - detects GST fusion proteins down to 7,5 ng/well
  • Flexibility - binds native or denatured forms of GST; an alternative to glutathione-coated plates
  • Ready to use - pre-blocked with SuperBlock Blocking Buffer
  • Compatibility - Thermo Scientific Cell Lysis Reagents and many other popular lysis buffers do not interfere with GST-fusion
More Product Information