Active GTPase pull-down and detection kits
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
16116 16117 16118 16119 16120 16121
PIER16116EA 1064 EUR
PIER16116 PIER16117 PIER16118 PIER16119 PIER16120 PIER16121
Active GTPase pull-down and detection kits
Testen Enzyme-assays
The Active GTPase Pull-Down and Detection Kits allows the monitoring of changes in GTPase activity levels by enriching and detecting the active (GTP-bound) state of a particular GTPase.
  • Validated – functionally tested to ensure quality and performance
  • Sensitive – optimised antibodies, reagents and Western blotting procedure accurately detect changes in GTPase activity levels
  • Convenient – kits contain controls and all reagents needed to perform and detect 30 pull-downs
  • Easy to use – conditions are optimised for immediate success in a 2-hour assay
  • Efficient – spin columns prevent sample loss
These kits include all the necessary reagents to lyse cells, enrich for active GTPases, positive and negative controls and an anti-GTPase primary antibody for Western blot detection.
Leveringsinformatie: Each kit includes GST fusion protein of specific binding domain (1 vial), glutathione agarose resin (3 ml), GTPγS, 100X (50 µl), GDP 100X (50 µl), lysis/binding/wash buffer (100 ml), GTPase-specific primary antibody (1 vial), SDS-PAGE sample loading buffer, 2X (1,5 ml), spin columns (30), and collection tubes (90).