Chemiluminescent EMSA kit, LightShift™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20148 20148X
PIER20148EA 852 EUR
PIER20148 PIER20148X
Chemiluminescent EMSA kit, LightShift™
Reagentia voor elektroforese Western Blotting Reagents
LightShift™ Chemiluminescent EMSA Kit is a robust and sensitive system for performing electrophoretic mobility shift assays (EMSA) to identify and characterise protein-DNA binding interactions.
  • Excellent for detecting low-abundance proteins in nuclear extracts
  • Sensitivity that surpasses radioactive and digoxigenin methods
  • Compatible with previously established binding conditions for popular DNA-protein interactions
  • Includes EBNA Optimisation and Control Kit to help new users develop a working assay and understand the methods used to confirm binding interaction specificity
Leveringsinformatie: Kit includes LightShift™ EMSA Optimisation and Control Kit (Cat. No. 20148X) and Chemiluminescent Nucleic Acid Detection Module (Cat. No. 89880).
Order Now
Specificaties
Environmentally Preferable