Immobilisation kit for proteins, SulfoLink™, Pierce™

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

20325 44995
PIER20325EA 291 EUR
PIER20325 PIER44995
Immobilisation kit for proteins, SulfoLink™, Pierce™
Eiwitzuivering Protein Purification Kits

SulfoLink™ Immobilisation Kit for Proteins is a kit containing all the reagents for preparing the protein sample for efficient immobilisation and columns containing SulfoLink™ resin that can be used in either gravity-flow or centrifuge format for efficient coupling reactions and multiple cycles of affinity purification.


  • Specific to sulphhydryl (-SH) groups
  • Prepare and couple protein samples in three and half hours
  • Flexible coupling conditions, use pH 7,5 to 9,0 aqueous buffers, organic solvent or denaturant
  • Streamlined protocols for sample preparation, immobilisation, and affinity purification
  • Immobilise 2 to 20 mg of protein per 2 ml column of SulfoLink™ coupling resin


When incubated with a solution of protein that contains reduced cysteine residues, the iodoacetyl groups of SulfoLink™ resin react specifically and efficiently with the exposed sulphhydryls (-SH) to form covalent and irreversible thioether bonds that permanently attach the protein to the resin. The result is a custom-made affinity resin for purification of antibodies, antigens and other molecules of interest.


Leveringsinformatie: Standard kit includes SulfoLink™ columns, 2 ml (5 columns), SulfoLink™ preparation buffer (7,5 ml), SulfoLink™ coupling buffer (500 ml), wash solution (120 ml), PBS pack (makes 500 ml), 2-Mercaptoethylamine·HCl (5×6 mg), L-Cysteine·HCl (100 mg), Zeba™ spin desalting columns, 5 ml (5 columns), and column accessories. A 1-column trial kit is also available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR