BCA™ protein assay kits and reagents

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

23208 23209 23223 23224 23210 23212 23230 23221 23213 23227 23228 23222 23225
PIER23208EA 195 EUR
PIER23208 PIER23209 PIER23223 PIER23224 PIER23210 PIER23212 PIER23230 PIER23221 PIER23213 786-0000 PIER23228 PIER23222 PIER23225
BCA™ protein assay kits and reagents
Testen Proteïne-assays

Pierce BCA™ (bicinchoninic acid) reagents provide accurate determination of protein concentration with most sample types encountered in protein research. The Pierce BCA™ Protein Assay can be used to assess yields in whole cell lysates and affinity-column fractions, as well as to monitor protein contamination in industrial applications. Compared to most dye-binding methods, the BCA™ Assay is affected much less by protein compositional differences, providing greater protein-to-protein uniformity.


  • Colorimetric, so can measure with a standard spectrophotometer or plate reader (562 nm)
  • Excellent uniformity, exhibiting less protein-to-protein variation than dye-binding methods
  • Unaffected by typical concentrations of most ionic and nonionic detergents
  • Easier and faster than the classical Lowry method
  • High linearity and sensitivity - linear working range for BSA 20 to 2000 µg/ml; detection down to 5 µg/ml (with the enhanced protocol)


BCA™ Protein Assay applications include studying protein:protein interactions; measuring column fractions after affinity chromatography; estimating percent recovery of membrane proteins from cell extracts; and high-throughput screening of fusion proteins.


Leveringsinformatie: Each kit contains BCA Reagent A (500 ml or 2×500 ml), BCA Reagent B (25 ml), and Albumin Standard Ampoules (2 mg/ml, 10×1 ml).

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR