Triethylamine

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: (Diethylamino)ethane, TEA

25108
PIER25108EA 193 EUR
PIER25108
Triethylamine
Triethylamine
Formule: (C₂H₅)₃N
MW: 101.19 g/mol
Kookpunt: 90
Smeltpunt: –115 °C
Dichtheid: 0.729 g/cm³ (20 °C)
Vlampunt: –11 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009051
CAS nummer: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3 II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR