5 mg, tablets

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 4-Nitrophenyl phosphate disodium salt hexahydrate, Phosphatase substrate, p-Nitrophenyl phosphate disodium salt hexahydrate, Phosphoric acid 4-nitrophenyl ester disodium salt hexahydrate, 4-Nitrophenyl phosphate disodium hexahydrate, pNPP disodium salt hexahydrate, di-Sodium 4-nitrophenyl phosphate hexahydrate

34047
PIER34047EA 221 EUR
PIER34047
5 mg, tablets
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP)
Formule: C₆H₄NNa₂O₆P·6H₂O
MW: 371.144 g/mol
Smeltpunt: 300
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00066288
CAS nummer: 4264-83-9
EINECS: 224-246-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR