Immobilised Protein G, UltraLink™, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
53125 53128 53126 53127
PIER53125EA 468 EUR
PIER53125 PIER53128 PIER53126 PIER53127
Immobilised Protein G, UltraLink™, Pierce™
Eiwitzuivering Protein Purification Resins
Pierce™ Protein G UltraLink™ and Protein G Plus UltraLink™ resins are useful for purification of antibodies and immunoprecipitation (IP). Samples containing IgG are incubated with the resin-immobilised Protein G in a buffer that facilitates binding. After non-IgG and non-antigen components of the sample are washed from the resin, the bound IgG and antigen may be recovered by elution.
  • Immobilised Protein G specifically binds IgG from several mammalian species, especially all isotypes of IgG from human, mouse and rat
  • Acrylamide based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
Protein G UltraLink™ resin binding capacity: >20 mg human IgG/ml resin.
Protein G UltraLink™ column binding capacity: >40 mg human IgG/ml resin.
Protein G Plus UltraLink™ resin binding capacity: >25 mg human IgG/ml resin.
Leveringsinformatie: Available as resin or as Protein G UltraLink Columns (2×2 ml resin pre-packed in columns with twist-off bottom tabs and an accessory pack containing two white tips).