Pierce™ Slide-A-Lyzer™ MINI Dialysis Devices

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

69550 69560 69552 69562 69572 69574 69576 69558 69580 69590 69553 69555 88404 88403 88400 88401 88405 88402
PIER69550EA 260 EUR
PIER69550 PIER69560 PIER69552 PIER69562 PIER69572 PIER69574 PIER69576 PIER69558 PIER69580 PIER69590 PIER69553 PIER69555 PIER88404 PIER88403 PIER88400 PIER88401 PIER88405 PIER88402
Pierce™ Slide-A-Lyzer™ MINI Dialysis Devices
Slangen Dialyseslangen
Slide-A-Lyzer MINI Dialysis devices, 3,5K MWCO, are disposable polypropylene cups with integrated, low-binding membranes for dialysis and high recovery of proteins and macromolecules >3,5 kDa in volumes from 10 µl to 2 ml. They were developed as a convenient means for dialysing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices.

  • Features of Slide-A-Lyzer MINI Dialysis Devices, 3,5K
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR