Affinity chromatography media, high capacity endotoxin removal resin, Pierce™

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

88270 88271 88272
PIER88270EA 419 EUR
PIER88270 PIER88271 PIER88272
Affinity chromatography media, high capacity endotoxin removal resin, Pierce™
Eiwitzuivering Protein Purification Resins

Pierce™ High Capacity Endotoxin Removal Resins are ideal for the removal of endotoxins from protein or antibody samples prior to cell transfection or animal injection.


  • Bind up to 2000000 EU/ml to eliminate <99% of endotoxins
  • Recover greater than 85% of protein sample


High Capacity Endotoxin Removal Resin is a porous cellulose bead that has been surface modified with covalently attached polylysine chains that have a high affinity for LPS but lacks the toxicity of other endotoxin-binding molecules. The resin in a slurry format helps in custom packing of endotoxin removal columns for gravity flow or flow rates ranging from 10 to 15 ml/hour. This can be packed to a custom column that can be re-used up to 10 times with no loss in endotoxin removal efficiency.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR