Chromatography cartridges, Protein A, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
89924 89925
PIER89924EA 279 EUR
PIER89924 PIER89925
Chromatography cartridges, Protein A, Pierce™
Kolommen voor chromatografie
Pierce™ Protein A chromatography cartridges are convenient, ready to use pre-packed devices for the isolation and purification of polyclonal and monoclonal antibodies. These cartridges are effective for routine affinity purification of polyclonal and monoclonal IgG from serum, ascites and cell culture supernatant.
  • Immobilised Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
  • Resin support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Superior
More Product Information