Chromatography cartridges, Protein A/G, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
89930 89931
PIER89930EA 348 EUR
PIER89930 PIER89931
Chromatography cartridges, Protein A/G, Pierce™
Kolommen voor chromatografie
Pierce™ Protein A/G chromatography cartridges are convenient, ready to use prepacked devices for isolation and purification of most immunoglobins.
  • Immobilised recombinant fusion protein of the antibody-binding domains of Protein A and Protein G enables polyclonal IgG purification from nearly any mammalian species
  • Resin support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Superior manufacturing method immobilises Protein A/G by charge-free, leach-resistant covalent bonds, resulting in low non
More Product Information