Deflected point septum penetration needles
Leverancier: CADENCE SCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4188
POPR4188EA 990 EUR
POPR4188
Deflected point septum penetration needles
Injectienaalden
These deflected needle style features point positioning on the axis of the needle lumen. This improved point geometry provides easier penetration and minimises coring and tracking (particularly with finer gauge needles).
Note: Needles not intended for human use.