Bottles, narrow neck, square or round, with screw top and thread
Leverancier: Poulten & Graf
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9.71004 9.70002 9.70004 9.70006
POUL9.71004EA 36.8 EUR
POUL9.71004 POUL9.70002 POUL9.70004 POUL9.70006
Bottles, narrow neck, square or round, with screw top and thread
Flessen Glazen flessen
Glass, amber.
  • Amber coloured to protect light sensitive substances.
Available coated or uncoated.