Cleaning trolley Clino®, CR mini EM-CR
Leverancier: PFENNIG REINIGUNGSTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3510150 3510140
PPSA3510150EA 923 EUR
PPSA3510150 PPSA3510140
Cleaning trolley Clino®, CR mini EM-CR
Transportwagens
The Clino® CR mini EM-CR model trolley system made of stainless steel suitable for cleanrooms and for non-sterile areas that require minimal space.
  • EasyMop GMP with ergonomically optimised system box
  • Highest efficiency and process reliability in single step technique
  • Basic module mini with high end finish and castors (100 mm)
  • EasyMop system boxes available in red, green, blue, yellow, grey and athracite
A small cleaning system with pre-preparation system EasyMop for technical cleanrooms suitable for ISO classes 7 and 8 or GMP areas C/D. An extraMore Product Information