Joint scrubber/brush for cleanrooms
Leverancier: PFENNIG REINIGUNGSTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3590600 3590510
PPSA3590600EA 38.5 EUR
PPSA3590600 PPSA3590510
Joint scrubber/brush for cleanrooms
Borstels
Joint scrubber/brush for gaps in hygienic sensitive areas.
  • Material: PP (body), PBT (stock)
  • Unslit or spouted fibre ends
  • Medium-hard bristle material
  • Heat-resistant up to 140 °C
Colour: White