Potassium chloride

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Potassiumchloride

BCR-113
PROOBCR-113EA 0 EUR
PROOBCR-113
Potassium chloride
Potassium chloride
Formule: KCl
Kookpunt: 1420 °C (1013 hPa)
Smeltpunt: 773 °C
Dichtheid: 1,98 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011360
CAS nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR