O-Acetylsalicylic acid

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Acetoxybenzoic acid, Acetylsalicylic acid, Aspirin, Aspirin®

BP617
PROOBP617EA 0 EUR
PROOBP617
O-Acetylsalicylic acid
O-Acetylsalicylic acid
Formule: 2-(CH₃CO₂)C₆H₄CO₂H
MW: 180.16 g/mol
Kookpunt: 140
Smeltpunt: 138…140 °C
Dichtheid: 1.39 g/cm³ (20 °C)
Vlampunt: 250
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002430
CAS nummer: 50-78-2
EINECS: 200-064-1
Merck Index: 13,00856
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fcb60a4ee505b596cef38216a85e034985091e0&vid=5f8d5cfeb3039783869a9320ec840b8ec12c33ea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR