Organic reference standards, Quinine sulphate standard, BP

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BP862
PROOBP862EA 0 EUR
PROOBP862
Organic reference standards, Quinine sulphate standard, BP
Standaarden Organische standaarden

Order Now

Specificaties

Omschrijving Quinine sulphate
Environmentally Preferable
CAS No. 6119-70-6
Formule C₂₀H₂₆N₂O₆S

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR