2-Methyl-1-propaanol

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol

CHE152
PROOCHE152EA 0 EUR
PROOCHE152
2-Methyl-1-propaanol
2-Methyl-1-propaanol
Formule: (CH₃)₂CHCH₂OH
MW: 74.12 g/mol
Kookpunt: 107.9
Smeltpunt: –108 °C
Dichtheid: 0.802 g/cm³ (20 °C)
Vlampunt: 28
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004740
CAS nummer: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3 III
Merck Index: 13,05148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR