2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110)

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CIL-PCB-110-C
PROOCIL-PCB-110-CEA 0 EUR
PROOCIL-PCB-110-C
2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110)
2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110)
Formule: C₁₂H₅Cl₅
MW: 326.44 g/mol
CAS nummer: 38380-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR