Amphotericin B

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

ICRS0210
PROOICRS0210EA 0 EUR
PROOICRS0210
Amphotericin B
Amphotericin B
Formule: C₄₇H₇₃NO₁₇
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00877763
CAS nummer: 1397-89-3
EINECS: 215-742-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR