3-Bromoanisole

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

MM0007.05
PROOMM0007.05EA 0 EUR
PROOMM0007.05
3-Bromoanisole
3-Bromoanisole
Formule: C₇H₇BrO
Kookpunt: 211 °C (1013 hPa)
Smeltpunt: 2 °C
Dichtheid: 1,49 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000081
CAS nummer: 2398-37-0
EINECS: 219-264-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR