1-Naftol

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene

MM0043.04
PROOMM0043.04EA 0 EUR
PROOMM0043.04
1-Naftol
1-Naftol
Formule: C₁₀H₈O
MW: 144.17 g/mol
Kookpunt: 282
Smeltpunt: 94
Dichtheid: 1.224 g/cm³ (20 °C)
Vlampunt: 125
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003930
CAS nummer: 90-15-3
EINECS: 201-969-4
UN: 2811
ADR: 6.1 III
Merck Index: 13,06409

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR