α,α,α-Trifluoro-p-cresol

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

MM0256.05
PROOMM0256.05EA 0 EUR
PROOMM0256.05
α,α,α-Trifluoro-p-cresol
α,α,α-Trifluoro-p-cresol
Formule: C₇H₅F₃O
Kookpunt: 185 °C (1013 hPa)
Smeltpunt: 45…47 °C
Dichtheid: 1,3 g/cm³ (25 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002363
CAS nummer: 402-45-9
EINECS: 206-945-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR