L-Alanine

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine

MM0566.00
PROOMM0566.00EA 0 EUR
PROOMM0566.00
L-Alanine
L-Alanine
Formule: C₃H₇NO₂
Kookpunt: 189 °C (1013 hPa)
Smeltpunt: 79 °C
Dichtheid: 1,4 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064410
CAS nummer: 56-41-7
EINECS: 200-273-8
Merck Index: 13,00203

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR