Saccharin sodium

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

MM0818.00
PROOMM0818.00EA 0 EUR
PROOMM0818.00
Saccharin sodium
Saccharin sodium
Formule: C₇H₅NNaO₃S
MDL Nummer: MFCD00013092
CAS nummer: 128-44-9
EINECS: 204-886-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR