Organische referentiestandaard, Dimetridazole 10 µg/ml in methanol

Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NE9245
PROONE9245EA 0 EUR
PROONE9245
Organische referentiestandaard, Dimetridazole 10 µg/ml in methanol
Standaarden Organische standaarden

Order Now

Specificaties

Omschrijving Dimetridazole
Environmentally Preferable
CAS No. 551-92-8
Formule C₅H₇N₃O₂

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR