Calcinated petroleum coke - trace elements
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
NIST-2719
PROONIST-2719EA 0 EUR
PROONIST-2719
Calcinated petroleum coke - trace elements
Standaarden Standaarden voor de petroleumindustrie
This reference standard is intended primarily for use in the calibration of apparatus and the evaluation of techniques employed in the analysis of calcined petroleum coke and other materials with a similar matrix.
Certificaten: Traceable to NIST and SRM numbers.