Nitrate ion chromatography standard, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-I1PNO3-100 VHG-INO3-100 VHG-INO3-500
PROOVHG-I1PNO3-100EA 0 EUR
PROOVHG-I1PNO3-100 PROOVHG-INO3-100 PROOVHG-INO3-500
Nitrate ion chromatography standard, VHG
Standaarden Anionische standaarden Nitraat standaarden
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certificaten: Traceable to NIST and SRM.
Order Now
Specificaties
Environmentally Preferable