Reference Materials, Matrix Modifiers for GFAA, CRM, VHG, Promochem
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-MMGN1P-100 VHG-MPM1-250 VHG-MPM2-250 VHG-MPM3-250
PROOVHG-MMGN1P-100EA 0 EUR
PROOVHG-MMGN1P-100 PROOVHG-MPM1-250 PROOVHG-MPM2-250 PROOVHG-MPM3-250
Reference Materials, Matrix Modifiers for GFAA, CRM, VHG, Promochem
Standaarden Kationische standaarden Standaarden met meerdere elementen
This solution is intended for use as a matrix modifier for graphite furnace atomic absorption spectroscopy (GFAA) in the determination of trace elements.
Certificaten: Traceable to NIST, SRM.