Antimony standard solution, for ICP, in 30% HCl, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PSBH-100 VHG-TSBH-100 VHG-PSBH-50 VHG-PSBH-500 VHG-TSBH-500
PROOVHG-PSBH-100EA 0 EUR
PROOVHG-PSBH-100 PROOVHG-TSBH-100 PROOVHG-PSBH-50 PROOVHG-PSBH-500 PROOVHG-TSBH-500
Antimony standard solution, for ICP, in 30% HCl, VHG
Standaarden Kationische standaarden Antimonium standaarden
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.