Strontium standard solution, for ICP, in HNO₃, CRM, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PSRN-100 VHG-PSRN-500 VHG-LSRN-100 VHG-PSRN-50 VHG-TSRN-100 VHG-TSRN-500
PROOVHG-PSRN-100EA 0 EUR
PROOVHG-PSRN-100 PROOVHG-PSRN-500 PROOVHG-LSRN-100 PROOVHG-PSRN-50 PROOVHG-TSRN-100 PROOVHG-TSRN-500
Strontium standard solution, for ICP, in HNO₃, CRM, VHG
Standaarden Kationische standaarden Strontium standaarden
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.