Tellurium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PTEN-100 VHG-PTEN-500
PROOVHG-PTEN-100EA 0 EUR
PROOVHG-PTEN-100 PROOVHG-PTEN-500
Tellurium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standaarden Kationische standaarden Tellurium standaarden
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.