Chromium standard solution, for ICP, in 5% HCl, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PCRH-100 VHG-PCRH-500 VHG-TCRH-100 VHG-TCRH-500
PROOVHG-PCRH-100EA 0 EUR
PROOVHG-PCRH-100 PROOVHG-PCRH-500 PROOVHG-TCRH-100 PROOVHG-TCRH-500
Chromium standard solution, for ICP, in 5% HCl, VHG
Standaarden Kationische standaarden Chroom standaarden
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.