Gadolinium standard solution, for ICP, in 5% HNO₃, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PGDN-100 VHG-PGDN-500 VHG-TGDN-100 VHG-TGDN-500
PROOVHG-PGDN-100EA 0 EUR
PROOVHG-PGDN-100 PROOVHG-PGDN-500 PROOVHG-TGDN-100 PROOVHG-TGDN-500
Gadolinium standard solution, for ICP, in 5% HNO₃, VHG
Standaarden Kationische standaarden Gadolinium standaarden
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.