Titanium standard solution, for ICP, in HNO₃, CRM, VHG
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHG-PTINF-100 VHG-LTINF-100 VHG-PTINF-50 VHG-PTINF-500 VHG-TTINF-100 VHG-TTINF-500
PROOVHG-PTINF-100EA 0 EUR
PROOVHG-PTINF-100 PROOVHG-LTINF-100 PROOVHG-PTINF-50 PROOVHG-PTINF-500 PROOVHG-TTINF-100 PROOVHG-TTINF-500
Titanium standard solution, for ICP, in HNO₃, CRM, VHG
Standaarden Kationische standaarden Titanium standaarden
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaten: Traceable to NIST, SRM.