Short-term storage solutions, 2-8CELLsius
Leverancier: PROTIDE PHARMACEUTICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PP338-100 PP338-500 PP338-1000 PP338-1000B
PRPHPP338-100EA 0 EUR
PRPHPP338-100 PRPHPP338-500 PRPHPP338-1000 PRPHPP338-1000B
Short-term storage solutions, 2-8CELLsius
Celcultuursupplementen en -additieven
Ready to use, balanced, cytoprotective, protein-free solution with bio-stabilisers for short-term storage, regenerative medicine, stem cell processing and transportation of biological material. Short-term storage can be achieved at 2 to 8 °C or at ambient temperature for 24 to 48 hours. Does not contain any growth factors, hormones, or antibiotics. 2-8CELLsius™ can also be utilised as a cell washing solution.
  • TYPE II drug master file
  • USP grade components/highest grade
  • Supports high cell viability
  • No animal or human proteins
  • No growth factors, hormones or antibiotics
  • Ready to use/sterile membrane filtered
Certificaten: cGMP manufactured.