Anti-FLIP Rabbit Polyclonal Antibody
Leverancier: ProSci Inc.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1156
PRSI1156EA 435 EUR
PRSI1156
Anti-FLIP Rabbit Polyclonal Antibody
Antilichamen
FLIP Antibody: Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain (DD) containing adapter molecules and members of the ICE/CED-3 protease family.Caspases-8 (FLICE) and -10 (FLICE2) are two pivotal members in the ICE/CED-3 protease family. FLICE-inhibitory proteins were identified in virus and human and designated v-FLIPs and FLIP respectively. The human FLIP was also cloned by several labs independently and termed Casper, I-FLICE, FLAME-1, CASH, CLARP and MRIT. FLIP contains two deathMore Product Information