Anti-KRAS Rabbit Polyclonal Antibody

Leverancier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14-589
PRSI14-589EA 519 EUR
PRSI14-589
Anti-KRAS Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: KRAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
New Product
Order Now

Specificaties

antigeen symbool KRAS
Type antilichaam Primair,Kirsten rat sarcoma viral oncogene homolog
Environmentally Preferable
Antigeen synoniemen KI-RAS|KRAS2|KRAS1|K-RAS2B|NS3|K-RAS4B|NS|RASK2|C-K-RAS|K-RAS4A|K-RAS2A|KRAS|CFC2
Type antilichaam Primair,Kirsten rat sarcoma viral oncogene homolog
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Rat,Mouse
Host Rabbit
Gen-ID 3845
Isotype IgG
Western blot Yes
Modification None
Immunogeen Recombinant fusion protein containing a sequence corresponding to amino acids 30-188 of human KRAS (NP_004976.2)
Formulier Liquid
Purification Affinity purification
Concentratie Batch dependent
Opslagbuffer PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
Opslagtemperatuur Store at –20...+730 °C. Avoid freeze/thaw cycles.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR