Anti-PCYT2 Rabbit Polyclonal Antibody

Leverancier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

15-980
PRSI15-980EA 419 EUR
PRSI15-980
Anti-PCYT2 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: PCYT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR