Anti-BCO2 Rabbit Polyclonal Antibody

Leverancier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

19-523
PRSI19-523EA 419 EUR
PRSI19-523
Anti-BCO2 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: BCO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
New Product
Order Now

Specificaties

antigeen symbool BCO2
Environmentally Preferable
Type antilichaam Primair,Beta,beta-carotene 9',10'-oxygenase
Antigeen synoniemen Beta-carotene dioxygenase 2|B-diox-II|Beta,beta-carotene 9',10'-oxygenase|BCDO2|BCO2
Type antilichaam Primair,Beta,beta-carotene 9',10'-oxygenase
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity
Host Rabbit
Gen-ID 83875
Isotype IgG
Western blot Yes
Immunogeen Recombinant fusion protein containing a sequence corresponding to amino acids 170-410 of human BCO2 (NP_001243327.1).
Formulier Liquid
Purification Affinity purification
Concentratie Batch dependent
Opslagbuffer PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
Opslagtemperatuur Store at −20...+730 °C. Avoid freeze/thaw cycles.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR