Anti-CEP120 Rabbit Polyclonal Antibody

Leverancier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

23-344
PRSI23-344EA 419 EUR
PRSI23-344
Anti-CEP120 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: CEP120
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR