Anti-TBXA2R Rabbit Polyclonal Antibody

Leverancier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

23-597
PRSI23-597EA 419 EUR
PRSI23-597
Anti-TBXA2R Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: TBXA2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR