Anti-GZMK Rabbit Polyclonal Antibody
Leverancier: ProSci Inc.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30-051
PRSI30-051EA 660 EUR
PRSI30-051
Anti-GZMK Rabbit Polyclonal Antibody
Antilichamen
GZMK is a member of a group of related serine proteases from the cytoplasmic granules of cytotoxic lymphocytes. Cytolytic T lymphocytes (CTL) and natural killer (NK) cells share the remarkable ability to recognize, bind, and lyse specific target cells. They are thought to protect their host by lysing cells bearing on their surface 'nonself' antigens, usually peptides or proteins resulting from infection by intracellular pathogens. The protein described here lacks consensus sequences for N-glycosylation present in other granzymes.This gene product is a member of a group of related serineMore Product Information