Anti-CD8A Mouse Monoclonal Antibody
Leverancier: ProSci Inc.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
32-134
PRSI32-134EA 390 EUR
PRSI32-134
Anti-CD8A Mouse Monoclonal Antibody
Antilichamen
CD8 T cell surface antigen is heterodimer of an alpha and a beta chain linked by two disulfide bonds.It belongs type I membrane protein. Selectively expressing of CD8 on a subset of T cells leads to CD8 T cell development. Through identifying cytotoxic/suppressor T-cells that interact with MHC class I bearing targets, CD8 is thought to play a role in the process of T-cell mediated killing. Veillette et al (1988) found the CD8 is associated with the internal membrane tyrosine-protein kinase p56lck.
Antibody is purified by protein A affinity chromatography.
Purified antibody in PBSMore Product Information