Anti-KRT7 Rabbit Monoclonal Antibody [clone: KRT7/1499R]
Leverancier: ProSci Inc.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
34-076
PRSI34-076EA 773 EUR
PRSI34-076
Anti-KRT7 Rabbit Monoclonal Antibody [clone: KRT7/1499R]
Antilichamen
Cytokeratin-7 (CK7) is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. CK7 is specifically expressed in the simple epithelia lining the cavities of the internal organs and in the gland ducts and blood vessels. It is found in simple glandular epithelia, and in transitional epithelium. Epithelial cells of the lung and breast both contain CK7, but some other glandular epithelia, such as those of the colon andMore Product Information