Anti-PIGO Rabbit Polyclonal Antibody

Leverancier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

59-078
PRSI59-078EA 620 EUR
PRSI59-078
Anti-PIGO Rabbit Polyclonal Antibody
Antilichamen
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: PIGO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 955-983
Host: Rabbit
Isotype: Ig
Reactivity: Human
New Product
Order Now

Specificaties

antigeen symbool PIGO
Environmentally Preferable
Type antilichaam Primair,GPI ethanolamine phosphate transferase 3
Type antilichaam Primair,GPI ethanolamine phosphate transferase 3
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Gen-ID 84720
Isotype
Western blot Yes
Modification None
Epitoop 955 - 983 aa
Immunogeen This PIGO antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 955 - 983 amino acids from the C-terminal region of human PIGO.
Formulier Liquid
Moleculair gewicht 119 kDa
Purification This antibody is purified through a protein A column, followed by peptide affinity purification
Concentratie 0,5 mg/ml
Opslagtemperatuur Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR