Anti-KPNA6 Rabbit Polyclonal Antibody

Leverancier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

5991
PRSI5991EA 435 EUR
PRSI5991
Anti-KPNA6 Rabbit Polyclonal Antibody
Antilichamen
KPNA6 Antibody: Karyopherin, a cytosolic and heterodimeric protein complex consisting of alpha and beta subunits, is responsible for targeting proteins with nuclear localization signals to the nuclear pore complex (NPC) by an energy requiring, Ran-dependent mechanism. The alpha subunit and imported substrate enter the nucleus and accumulate in the nucleoplasm, while the beta subunit accumulates at the NPC. KPNA6 belongs to a subfamily within the KPNA family that also includes IPOA5&6. Down-regulation of KPNA6 by RNAi expression in HeLa cells strongly inhibited cell proliferation,More Product Information
Order Now

Specificaties

antigeen symbool KPNA6
Type antilichaam Karyopherin Alpha 6
Environmentally Preferable
Antigeen synoniemen KPNA6 |KPNA7|FLJ11249|importin-alpha-S2|IPOA7|importin alpha 7 subunit|karyopherin alpha 6 (importin alpha 7) |MGC17918|karyopherin alpha 6
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoFluorescentie Yes
Immunogeen KPNA6 antibody was raised against a 13 amino acid synthetic peptide near the amino terminus of human KPNA6.
Purification Affinity chromatography purified via peptide column
Opslagbuffer PBS containing 0.02% sodium azide
Tested applications E, WB, IF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR